Assay Details
| Gene Symbol: Gm10248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033550.1 | 2 - 3 | 712 | 75 | ||
| GenBank mRNA | AY512910.1 | - | 2 - 3 | 712 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700112E06 gene | 1700112E06Rik | 2700009F18Rik | Anti-sense | Chr. 14 - 22019503 - 23056088 on NCBI Build GRCm38 | |
| Downstream |
potassium large conductance calcium-activated channel, subfamily M, alpha member 1 |
Kcnma1 |
5730414M22Rik, BKCa, MaxiK, Slo, Slo1, mSlo, mSlo1 |
198.6kb | Sense | Chr. 14 - 23292291 - 24004951 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY512910.1 transcripts.
Set Membership: |
|

Gm10248
Genomic Map