Assay Details
| Gene Symbol: Gm10248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033550.1 | 3 - 4 | 778 | 117 | ||
| GenBank mRNA | AY512910.1 | - | 3 - 4 | 778 | 117 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700112E06 gene | 1700112E06Rik | 2700009F18Rik | Anti-sense | Chr. 14 - 22019503 - 23056088 on NCBI Build GRCm38 | |
| Downstream |
potassium large conductance calcium-activated channel, subfamily M, alpha member 1 |
Kcnma1 |
5730414M22Rik, BKCa, MaxiK, Slo, Slo1, mSlo, mSlo1 |
251.1kb | Sense | Chr. 14 - 23292291 - 24004951 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY512910.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P208325 | AY512910.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P208325 | AY512910.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158977 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P180042 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08052591 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158977 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P180042 | - | - |
| Whole Mouse Genome G4122A | A_52_P208325 | AY512910.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P208325 | AY512910.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117917 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158977 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P180042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11434639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462940 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462990 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11348873 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462871 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11213454 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11252889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462694 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462923 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462878 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462916 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462906 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11786740 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462747 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11741253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462974 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462790 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11663582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462796 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462715 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462726 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463052 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11509057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11348869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11194821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11741232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463075 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462779 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462863 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462987 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11741233 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462963 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462972 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462989 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11741255 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462823 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11241653 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11285636 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11291020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462961 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11207107 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11805815 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462773 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462922 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11741236 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11771718 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462800 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11674065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11162941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11786738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11786744 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462860 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11662382 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462824 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11771709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11296678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11805812 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11348854 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462703 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462762 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462771 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11296688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11296687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462693 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11741235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11348864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11252898 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462730 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11555823 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462802 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462827 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462728 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462740 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11674050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11502947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11771720 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462867 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462809 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462978 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462854 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462718 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462720 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462781 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11555826 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462957 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462697 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462710 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11272258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11285634 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462849 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462842 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462759 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462969 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11292189 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462830 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462905 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462749 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462844 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462702 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463082 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462921 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462845 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11741246 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462774 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462851 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11741222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11209872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462808 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11738202 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11296684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11290666 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462914 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462712 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462865 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11162943 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462797 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462723 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462731 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462769 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462992 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11181462 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462705 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462793 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11559986 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462857 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11213451 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462982 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29463068 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29462909 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.1 ST Array | 10418048 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400011774.mm.1 | NR_033550.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400011774.mm.1 | NR_033550.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400011774.mm.1 | TC1400001566.mm.1 | MTA TC |

Gm10248
Genomic Map