Assay Details
Gene Symbol: Gm10280 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033584.1 | 1 - 2 | 93 | 116 | ||
GenBank mRNA | AY512935.1 | - | 1 - 2 | 93 | 116 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | contactin associated protein-like 4 | Cntnap4 |
Caspr4, E130114F09Rik |
-184.6kb | Sense | Chr. 8 - 112569965 - 112882707 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 4930571O06 gene | 4930571O06Rik | 428.2kb | Sense | Chr. 8 - 113517282 - 113550808 on NCBI Build GRCm38 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AY512935.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
