Assay Details
| Gene Symbol: 4930503H13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033598.1 | 1 | 50 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | patched domain containing 1 | Ptchd1 |
9630036J22Rik, AI848149, Gm387 |
-915.7kb | Anti-sense | Chr. X - 155569736 - 155623812 on NCBI Build GRCm38 |
| Downstream |
phosphate regulating endopeptidase homolog, X-linked |
Phex |
Gy, HPDR, HPDR1, Hyp, PEX |
622.0kb | Anti-sense | Chr. X - 157162075 - 157416551 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930503H13Rik
Genomic Map