Assay Details
| Gene Symbol: Gm5176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033603.1 | 1 | 256 | 74 | ||
| GenBank mRNA | BC054110.1 | - | 319 | 30017181 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nucleosome assembly protein 1-like 1 | Nap1l1 |
AA407126, AI256722, D10Ertd68e, NAP-1 |
-2.6kb | Sense | Chr. 10 - 111473192 - 111498150 on NCBI Build GRCm38 |
| Downstream |
pleckstrin homology-like domain, family A, member 1 |
Phlda1 |
DT1P1B11, TDAG51, Tdag |
4.9kb | Sense | Chr. 10 - 111506286 - 111508649 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC054110.1 transcripts.
Set Membership: |
|

Gm5176
Genomic Map