Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1444177_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1444177_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1444177_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0100026652.mm.1 |
NR_033736.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026652.mm.1 |
NR_033736.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026652.mm.1 |
TC0100003276.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026645.mm.1 |
NR_033736.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026645.mm.1 |
NR_033736.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026645.mm.1 |
TC0100003276.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026643.mm.1 |
NR_033736.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026643.mm.1 |
NR_033736.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026643.mm.1 |
TC0100003276.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026651.mm.1 |
NR_033736.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026651.mm.1 |
NR_033736.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026651.mm.1 |
TC0100003276.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026647.mm.1 |
NR_033736.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026647.mm.1 |
NR_033736.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026647.mm.1 |
TC0100003276.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026648.mm.1 |
NR_033736.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026648.mm.1 |
NR_033736.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026648.mm.1 |
TC0100003276.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026653.mm.1 |
NR_033736.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026653.mm.1 |
NR_033736.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026653.mm.1 |
TC0100003276.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026644.mm.1 |
NR_033736.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026644.mm.1 |
NR_033736.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026644.mm.1 |
TC0100003276.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026646.mm.1 |
NR_033736.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026646.mm.1 |
NR_033736.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026646.mm.1 |
TC0100003276.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1444177_at |
- |
- |
Mouse Expression Set 430 |
1444177_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1444177_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1444177_at |
- |
- |