Assay Details
| Gene Symbol: Mir1933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_035454.1 | 1 | 40 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | vacuolar protein sorting 54 (yeast) | Vps54 |
Hcc8, Vps54l, mSLP8, wr |
-23.5kb | Anti-sense | Chr. 11 - 21239032 - 21321133 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4932414J04 gene | 4932414J04Rik | 150.1kb | Sense | Chr. 11 - 21494730 - 21511180 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay targets the same location as MTA transcript cluster(s) TC1100002325.mm
This assay targets the same location as RTA transcript cluster(s) TC0X00001592.rn
This assay targets the same location as RTA transcript cluster(s) TC0X00001592.rn
Set Membership: |
|

Mir1933
Genomic Map