Assay Details
| Gene Symbol: Snora74a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002905.3 | 1 | 47 | 92 | ||
| Gene Symbol: Snhg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK088959.1 | - | 3 | 555 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 1949 | Mir1949 |
mir-1949, mmu-mir-1949 |
-2.4kb | Sense | Chr. 18 - 35554567 - 35554636 on NCBI Build GRCm38 |
| Downstream | matrin 3 | Matr3 |
1110061A14Rik, 2810017I02Rik, AI841759, AW555618, D030046F20Rik, mKIAA0723 |
4.9kb | Sense | Chr. 18 - 35562158 - 35592045 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snora74a
Genomic Map