Assay Details
| Gene Symbol: Mirlet7c-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029728.1 | 1 | 49 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 31322 |
Gm31322 | Sense | Chr. 16 - 77589379 - 77790662 on NCBI Build GRCm38 | ||
| Downstream | microRNA 125b-2 | Mir125b-2 |
Mirn125b-2, mir-125b-2 |
46.5kb | Sense | Chr. 16 - 77646273 - 77646343 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mirlet7c-1
Genomic Map