Assay Details
| Gene Symbol: Scarna3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028518.1 | 1 | 55 | 92 | ||
| GenBank mRNA | AF357404.1 | - | 1 | 55 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
POU domain, class 5, transcription factor 1 pseudogene |
Gm21053 |
Oct4, Pou5f1 |
-46.1kb | Sense | Chr. 1 - 159293805 - 159294189 on NCBI Build GRCm38 |
| Downstream | tenascin R | Tnr |
J1-tenascin, TN-R, janusin, restrictin |
183.3kb | Sense | Chr. 1 - 159523741 - 159931729 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Scarna3a
Genomic Map