Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10541090 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10489044 |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1428286_PM_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1432646_PM_a_at |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10541089 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10489043 |
- |
- |
Mouse Genome 430 2.0 Array |
1428286_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1428286_at |
- |
- |
Mouse Genome 430 2.0 Array |
1432646_PM_a_at |
- |
- |
Mouse Genome 430 2.0 Array |
1432646_a_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0200042648.mm.1 |
NR_024329.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042648.mm.1 |
NR_024329.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042648.mm.1 |
TC0200004989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200042644.mm.1 |
NR_024329.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042644.mm.1 |
NR_024329.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042644.mm.1 |
TC0200004989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200042646.mm.1 |
NR_024329.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042646.mm.1 |
NR_024329.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042646.mm.1 |
TC0200004989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200042647.mm.1 |
NR_024329.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042647.mm.1 |
NR_024329.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042647.mm.1 |
TC0200004989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200042650.mm.1 |
NR_024329.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042650.mm.1 |
NR_024329.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042650.mm.1 |
TC0200004989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200042652.mm.1 |
NR_024329.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042652.mm.1 |
NR_024329.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042652.mm.1 |
TC0200004989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200042654.mm.1 |
NR_024329.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042654.mm.1 |
NR_024329.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042654.mm.1 |
TC0200004989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200042645.mm.1 |
NR_024329.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042645.mm.1 |
NR_024329.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042645.mm.1 |
TC0200004989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200042649.mm.1 |
NR_024329.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042649.mm.1 |
NR_024329.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042649.mm.1 |
TC0200004989.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1428286_at |
- |
- |
Mouse Expression Set 430 |
1432646_PM_a_at |
- |
- |
Mouse Expression Set 430 |
1428286_PM_at |
- |
- |
Mouse Expression Set 430 |
1432646_a_at |
- |
- |
Murine Genome U74v2 Set |
95848_at |
- |
- |
Murine Genome U74v2 Set |
96299_at |
- |
- |
Murine Genome U74v2 Set |
96299_PM_at |
- |
- |
Murine Genome U74v2 Set |
95848_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1432646_PM_a_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1428286_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1428286_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1432646_a_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1428286_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1432646_a_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1428286_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1432646_PM_a_at |
- |
- |