Assay Details
| Gene Symbol: D430020J02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028421.1 | 2 | 2653 | 104 | ||
| GenBank mRNA | AK084974.1 | - | 2 | 2628 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | extended synaptotagmin-like protein 2 | Esyt2 |
2410017M09Rik, 4921504I16Rik, CHR2SYT, D12Ertd551e, Fam62b |
-11.0kb | Anti-sense | Chr. 12 - 116279680 - 116390935 on NCBI Build GRCm38 |
| Downstream |
non-SMC condensin II complex, subunit G2 |
Ncapg2 |
5830426I05Rik, Luzp5, Mtb, mCAP-G2 |
714.0 | Anti-sense | Chr. 12 - 116405357 - 116463532 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

D430020J02Rik
Genomic Map