Assay Details
| Gene Symbol: Slc22a13b-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033303.1 | 6 | 1151 | 95 | ||
| GenBank mRNA | AK079093.1 | - | 6 | 1078 | 95 | |
| BC147282.1 | - | 6 | 875 | 9056356 | 95 | |
| BC147283.1 | - | 6 | 875 | 9056343 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 22 (organic cation transporter), member 13 |
Slc22a13 |
AI648912, OCTL1, OCTL3, ORCTL3 |
-11.4kb | Sense | Chr. 9 - 119192968 - 119209105 on NCBI Build GRCm38 |
| Downstream | oxidative-stress responsive 1 | Oxsr1 |
2210022N24Rik, 2810422B09Rik, AI462649, AW209236, Osr1, mKIAA1101 |
17.5kb | Sense | Chr. 9 - 119238432 - 119322710 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a13b-ps
Genomic Map