Assay Details
Gene Symbol: Gm1110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281475.1 | NP_001268404.1 | 18 - 19 | 1809 | 76 | |
GenBank mRNA | AK136683.1 | - | 9 - 10 | 1235 | 76 | |
DQ664197.1 | - | 18 - 19 | 1818 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
