Assay Details
Gene Symbol: Gm1110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281475.1 | NP_001268404.1 | 20 - 21 | 1989 | 115 | |
GenBank mRNA | AK136683.1 | - | 11 - 12 | 1415 | 115 | |
DQ664197.1 | - | 20 - 21 | 1998 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133511 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2107967 | DQ664197.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2107967 | NM_001177509.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01890077 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373123 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07866426 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373105 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373045 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373087 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07573959 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07525671 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373100 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373153 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07704309 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373091 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07698942 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07878515 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373094 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373070 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373042 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373149 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07935366 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07573960 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373072 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373083 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373133 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373058 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373141 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07573964 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373136 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373127 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373047 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373060 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373081 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373049 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373103 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07866425 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373084 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07545915 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07878513 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373130 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373071 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373158 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373074 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373106 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373125 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07545924 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07545918 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373121 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373102 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373050 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373062 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373041 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373040 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373068 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373138 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373066 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07704307 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373080 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373076 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26373132 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07716498 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10591934 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591936 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591937 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591946 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591928 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591929 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591941 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591933 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591944 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591943 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591931 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591942 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591935 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591939 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591940 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591938 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591932 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591930 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591945 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591927 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10591926 | - | - |
Affymetrix Mouse Clariom S Array | PSR0900016297.mm.1 | NM_001281475.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900016297.mm.1 | TC0900001941.mm.1 | MTA TC |
