Assay Details
Gene Symbol: Gps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177874.1 | NP_001171345.1 | 6 - 7 | 725 | 83 | |
NM_145370.2 | NP_663345.2 | 6 - 7 | 865 | 83 | ||
GenBank mRNA | AK088490.1 | - | 6 - 7 | 722 | 83 | |
BC003350.1 | - | 6 - 7 | 865 | 3481979 | 83 | |
BC103782.1 | - | 6 - 7 | 692 | 6486599 | 83 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
