Assay Details
Gene Symbol: Gps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177874.1 | NP_001171345.1 | 8 - 9 | 873 | 73 | |
NM_145370.2 | NP_663345.2 | 8 - 9 | 1013 | 73 | ||
GenBank mRNA | AK088490.1 | - | 8 - 9 | 870 | 73 | |
BC003350.1 | - | 8 - 9 | 1013 | 3481979 | 73 | |
BC103782.1 | - | 8 - 9 | 840 | 6486599 | 73 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28186730 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28186732 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28186733 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28186734 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2143968 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087385 | BC103782.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087385 | AK088490.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087385 | BC003350.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087385 | NM_145370.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087385 | NM_001177874.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087384 | BC103782.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087384 | BC003350.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087384 | NM_145370.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087384 | NM_001177874.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087384 | AK049333.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 140700 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 506197 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 52058 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149786 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087385 | BC103782.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087385 | AK088490.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087385 | BC003350.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087385 | NM_145370.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087385 | NM_001177874.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087384 | BC103782.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087384 | BC003350.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087384 | NM_145370.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087384 | NM_001177874.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087384 | AK049333.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2143968 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186754 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186745 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186757 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186767 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186737 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186734 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186740 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186749 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186761 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186759 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186762 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186739 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186746 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186766 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186730 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186756 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186742 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149786 | - | - |
Whole Mouse Genome G4122A | A_51_P374453 | BC103782.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P374453 | AK088490.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P374453 | BC003350.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P374453 | NM_145370.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P374453 | NM_001177874.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114780 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121771 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149786 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09736791 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09736770 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28186733 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28186734 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28186732 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28186730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186830 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186820 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186818 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186815 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186816 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186801 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186810 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186807 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186794 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186788 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186761 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28186739 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28186742 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28186761 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28186764 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28186752 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09736816 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28186746 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28186737 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28186740 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28186734 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28186757 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09736772 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09736789 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28186766 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28186756 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09736783 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09736815 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10383497 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10383486 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10383499 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10383498 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10383501 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10383496 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10383493 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10383500 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10436448 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10383490 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10383487 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10383488 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10383494 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10383489 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10436447 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10383491 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10383495 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10383492 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1415699_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10436446 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10383485 | - | - |
Mouse Genome 430 2.0 Array | 1415699_a_at | - | - |
Mouse Genome 430 2.0 Array | 1415699_PM_a_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100019794.mm.1 | NM_001177874.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019794.mm.1 | NM_001177874.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019794.mm.1 | NM_145370.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019794.mm.1 | NM_145370.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019794.mm.1 | TC1100002048.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100019789.mm.1 | NM_001177874.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019789.mm.1 | NM_001177874.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019789.mm.1 | NM_145370.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019789.mm.1 | NM_145370.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019789.mm.1 | TC1100002048.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100019790.mm.1 | NM_001177874.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019790.mm.1 | NM_001177874.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019790.mm.1 | NM_145370.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019790.mm.1 | NM_145370.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019790.mm.1 | TC1100002048.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100019792.mm.1 | NM_001177874.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019792.mm.1 | NM_001177874.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019792.mm.1 | NM_145370.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019792.mm.1 | NM_145370.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019792.mm.1 | TC1100002048.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100019791.mm.1 | NM_001177874.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019791.mm.1 | NM_001177874.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019791.mm.1 | NM_145370.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019791.mm.1 | NM_145370.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019791.mm.1 | TC1100002048.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100019785.mm.1 | NM_001177874.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019785.mm.1 | NM_001177874.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019785.mm.1 | NM_145370.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019785.mm.1 | NM_145370.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019785.mm.1 | TC1100002048.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100019795.mm.1 | NM_001177874.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019795.mm.1 | NM_001177874.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019795.mm.1 | NM_145370.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019795.mm.1 | NM_145370.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019795.mm.1 | TC1100002048.mm.1 | MTA TC |
Mouse Expression Set 430 | 1415699_PM_a_at | - | - |
Mouse Expression Set 430 | 1415699_a_at | - | - |
Mouse Expression Set 430 | 1415699_PM_a_at | - | - |
Mouse Expression Set 430 | 1415699_a_at | - | - |
Murine Genome U74v2 Set | 96943_at | - | - |
Murine Genome U74v2 Set | 96943_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1415699_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1415699_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1415699_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1415699_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1415699_a_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1415699_PM_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 901711 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2470037 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 7100040 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4760315 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2470037 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6200528 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0007100040 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002470037 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006200528 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004760315 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2632714 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2625982 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2601090 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1254788 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2632714 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2625982 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2601090 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1254788 | - | - |
mouse-6 v1.1 (Illumina) | scl00209318.1_9-S | - | - |
mouse-6 v1.1 (Illumina) | scl0209318.14_24-S | - | - |
mouse-6 v1.1 (Illumina) | scl0001480.1_10-S | - | - |
mouse-6 v1.1 (Illumina) | scl0209318.1_47-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 7100040 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4760315 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2470037 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6200528 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00209318.1_9-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0209318.14_24-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0209318.1_47-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001480.1_10-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4760315 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2470037 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7100040 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6200528 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2632714 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1254788 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2625982 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2601090 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2632714 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2601090 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2625982 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1254788 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0209318.14_24-S | - | - |
