Assay Details
Gene Symbol: Wdr53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185162.1 | NP_001172091.1 | 2 - 3 | 539 | 67 | |
NM_026898.2 | NP_081174.1 | 2 - 3 | 771 | 67 | ||
GenBank mRNA | AK005105.1 | - | 2 - 3 | 738 | 67 | |
BC028850.1 | - | 1 - 2 | 503 | 3987279 | 67 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30651594 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P219028 | NM_026898.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P219028 | AK005105.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P219028 | NM_001185162.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P219028 | BC028850.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P219028 | NM_026898.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P219028 | AK005105.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P219028 | NM_001185162.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P219028 | BC028850.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 275144 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 415636 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 364117 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 773948 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 455677 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 839512 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 665992 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 109601 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130295 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P219028 | NM_026898.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P219028 | AK005105.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P219028 | NM_001185162.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P219028 | BC028850.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P219028 | NM_026898.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P219028 | AK005105.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P219028 | NM_001185162.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P219028 | BC028850.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30651598 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30651601 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30651613 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30651625 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30651597 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30651594 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30651610 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30651624 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30651603 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30651604 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30651616 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30651602 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30651611 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30651609 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30651606 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30651618 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30651599 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30651615 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30651595 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P130295 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08554014 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08554035 | - | - |
Whole Mouse Genome G4122A | A_51_P219028 | NM_026898.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P219028 | AK005105.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P219028 | NM_001185162.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P219028 | BC028850.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P219028 | NM_026898.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P219028 | AK005105.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P219028 | NM_001185162.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P219028 | BC028850.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P130295 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12863972 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13042947 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30651594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30651599 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30651607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30651636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30651595 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30651613 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30651609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30651598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30651630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30651625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30651624 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30651652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30651611 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30651616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30651592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30651658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30651597 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30651601 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30651660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30651651 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30651654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30651656 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12863975 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30651625 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30651611 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30651631 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30651601 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30651603 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30651604 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30651615 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30651607 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30651613 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30651598 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30651627 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30651609 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30651630 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30651618 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30651616 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30651595 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30651624 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30651606 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30651614 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30651621 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30651610 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12847047 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30651628 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12863974 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10435018 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10435017 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10435016 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1428839_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1428840_PM_s_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10435015 | - | - |
Mouse Genome 430 2.0 Array | 1428840_s_at | - | - |
Mouse Genome 430 2.0 Array | 1428839_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1428840_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1428839_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68980_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68980_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1600004002.mm.1 | NM_001185162.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600004002.mm.1 | NM_001185162.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600004002.mm.1 | TC1600000434.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1600004011.mm.1 | NM_001185162.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600004011.mm.1 | NM_001185162.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600004011.mm.1 | NM_026898.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600004011.mm.1 | NM_026898.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600004011.mm.1 | TC1600000434.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1600004013.mm.1 | NM_001185162.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600004013.mm.1 | NM_001185162.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600004013.mm.1 | NM_026898.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600004013.mm.1 | NM_026898.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600004013.mm.1 | TC1600000434.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1600004012.mm.1 | NM_001185162.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600004012.mm.1 | NM_001185162.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600004012.mm.1 | NM_026898.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600004012.mm.1 | NM_026898.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600004012.mm.1 | TC1600000434.mm.1 | MTA TC |
Mouse Expression Set 430 | 1428839_at | - | - |
Mouse Expression Set 430 | 1428840_s_at | - | - |
Mouse Expression Set 430 | 1428839_PM_at | - | - |
Mouse Expression Set 430 | 1428840_PM_s_at | - | - |
Murine Genome U74v2 Set | 171102_i_at | - | - |
Murine Genome U74v2 Set | 171102_PM_i_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1428839_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1428840_s_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1428840_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1428839_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 807316 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2706964 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2706964 | - | - |
