Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100031612.mm.1 |
NM_001198955.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031612.mm.1 |
NM_001198955.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031612.mm.1 |
TC0100003886.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100028702.mm.1 |
NM_001198955.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100028702.mm.1 |
NM_001198955.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100028702.mm.1 |
TC0100003886.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100028699.mm.1 |
NM_001198955.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100028699.mm.1 |
NM_001198955.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100028699.mm.1 |
TC0100003886.mm.1 |
MTA TC |