Assay Details
| Gene Symbol: Srd5a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020611.4 | NP_065636.2 | 2 - 3 | 420 | 104 | |
| GenBank mRNA | AK040198.1 | - | 2 - 3 | 388 | 104 | |
| AK075635.1 | - | 2 - 3 | 416 | 104 | ||
| AK085748.1 | - | 2 - 3 | 269 | 104 | ||
| AK138681.1 | - | 2 - 3 | 412 | 104 | ||
| AK139290.1 | - | 2 - 3 | 417 | 104 | ||
| AK162527.1 | - | 2 - 3 | 388 | 104 | ||
| BC039564.1 | - | 2 - 3 | 373 | 104 | ||
| BC145647.1 | - | 2 - 3 | 40130950 | 104 | ||
More Information
Set Membership: |
|

Srd5a3
Genomic Map