Assay Details
| Gene Symbol: Srd5a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020611.4 | NP_065636.2 | 4 - 5 | 654 | 65 | |
| GenBank mRNA | AK040198.1 | - | 4 - 5 | 622 | 65 | |
| AK075635.1 | - | 4 - 5 | 650 | 65 | ||
| AK085748.1 | - | 4 - 5 | 503 | 65 | ||
| AK139290.1 | - | 4 - 5 | 651 | 65 | ||
| AK162527.1 | - | 4 - 5 | 622 | 65 | ||
| BC039564.1 | - | 4 - 5 | 607 | 65 | ||
More Information
Set Membership: |
|

Srd5a3
Genomic Map