Assay Details
| Gene Symbol: Tmem254a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025311.3 | NP_079587.1 | 1282 | 93 | ||
| NM_026679.2 | NP_080955.1 | 1451 | 93 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29480509 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29480508 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29480510 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29480513 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29480511 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967553 | AK003554.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967553 | AK008299.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967553 | BC031854.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967553 | NM_025311.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967553 | NM_026679.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2110290 | AV008778.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2110290 | NM_025311.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2110290 | NM_026679.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967553 | AK003554.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967553 | AK008299.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967553 | BC031854.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967553 | NM_025311.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967553 | NM_026679.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480531 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29480500 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02815851 | - | - |
| Whole Mouse Genome G4122A | A_51_P222453 | AK003554.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P222453 | AK008299.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P222453 | BC031854.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P222453 | NM_025311.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P222453 | NM_026679.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114772 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02815851 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11721186 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11390010 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11721190 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11721193 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11316105 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11726090 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11721185 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29480513 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29480508 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29480509 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29480511 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29480510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29480544 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480478 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11781988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480542 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11781990 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480476 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480517 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480497 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480495 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480466 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480469 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480504 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480473 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480509 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480528 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480488 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11438500 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480531 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480490 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480470 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11815667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480516 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11324690 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480537 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480527 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480544 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480541 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11300882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11356759 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480481 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480501 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480479 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480494 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480505 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480491 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11721183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11580918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480499 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11405910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480510 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480489 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480468 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11341381 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480506 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480519 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480500 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11324691 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29480536 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10418204 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10418202 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10418215 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10418192 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10418212 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10418199 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10418188 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10418214 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10418191 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10418189 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10418213 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10418201 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10418187 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10418190 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10418203 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10418216 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10418200 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428738_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10418210 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10418185 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10418198 | - | - |
| Mouse Genome 430 2.0 Array | 1428738_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1428738_PM_a_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400012071.mm.1 | NM_001270499.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400012071.mm.1 | NM_025311.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400012071.mm.1 | NM_025311.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400012071.mm.1 | NM_026679.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400012071.mm.1 | NM_026679.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400012071.mm.1 | TC1400001605.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400012084.mm.1 | NM_026679.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400012084.mm.1 | NM_026679.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400012084.mm.1 | TC1400001605.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400012085.mm.1 | NM_026679.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400012085.mm.1 | NM_026679.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400012085.mm.1 | TC1400001605.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400012068.mm.1 | NM_025311.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400012068.mm.1 | NM_025311.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400012068.mm.1 | NM_026679.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400012068.mm.1 | NM_026679.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400012068.mm.1 | TC1400001605.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428738_a_at | - | - |
| Mouse Expression Set 430 | 1428738_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100360397 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1340440 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100360397 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001340440 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2693858 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1258879 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2693858 | - | - |
| mouse-6 v1.1 (Illumina) | scl45800.5_19-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0066039.2_47-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1340440 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100360397 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0066039.2_47-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100360397 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1340440 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2693858 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2693858 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0066039.2_47-S | - | - |

Tmem254a
Genomic Map