Assay Details
| Gene Symbol: 4933406F09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015568.2 | 1 - 2 | 134 | 105 | ||
| GenBank mRNA | AK005712.1 | - | 127 | 105 | ||
| AK015236.1 | - | 148 | 105 | |||
| AK016686.1 | - | 1 - 2 | 136 | 105 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4930521O11 gene | 4930521O11Rik | -30.0 | Anti-sense | Chr. 14 - 7362097 - 7412443 on NCBI Build GRCm38 | |
| Downstream | predicted gene 3636 | Gm3636 | 100042043 | 375.4kb | Anti-sense | Chr. 14 - 6736742 - 6757604 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4933406F09Rik
Genomic Map