Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700001494.mm.1 |
TC0700000214.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700001494.mm.1 |
NR_038092.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001494.mm.1 |
NR_038092.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001493.mm.1 |
TC0700000214.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700001493.mm.1 |
NR_038092.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001493.mm.1 |
NR_038092.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001495.mm.1 |
TC0700000214.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700001495.mm.1 |
NR_038092.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001495.mm.1 |
NR_038092.1 |
Refseq |