Assay Details
| Gene Symbol: Plxna4os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040277.1 | 1 - 2 | 311 | 88 | ||
| GenBank mRNA | AK038471.1 | - | 1 - 2 | 307 | 88 | |
| AK046388.1 | - | 1 - 2 | 225 | 88 | ||
| BB535018.2 | - | 1 - 2 | 311 | 88 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | plexin A4 | Plxna4 |
9330117B14, Plxa4, mKIAA1550 |
Anti-sense | Chr. 6 - 32144543 - 32588192 on NCBI Build GRCm38 | |
| Downstream | coiled-coil-helix-coiled-coil-helix domain containing 3 | Chchd3 |
0610041L09Rik, 1700039J09Rik, AW558177 |
276.6kb | Anti-sense | Chr. 6 - 32792217 - 33060276 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Plxna4os1
Genomic Map