Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700009910.mm.1 |
TC0700001124.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009910.mm.1 |
NR_040311.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009910.mm.1 |
NR_040311.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009910.mm.1 |
NR_040312.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009910.mm.1 |
NR_040312.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009910.mm.1 |
NR_040314.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009910.mm.1 |
NR_040314.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009912.mm.1 |
TC0700001124.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009912.mm.1 |
NR_040311.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009912.mm.1 |
NR_040311.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009903.mm.1 |
TC0700001124.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009903.mm.1 |
NR_040311.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009903.mm.1 |
NR_040311.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009903.mm.1 |
NR_040313.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009903.mm.1 |
NR_040313.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009903.mm.1 |
NR_040314.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009903.mm.1 |
NR_040314.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009905.mm.1 |
TC0700001124.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009905.mm.1 |
NR_040311.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009905.mm.1 |
NR_040311.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009905.mm.1 |
NR_040313.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009905.mm.1 |
NR_040313.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009905.mm.1 |
NR_040314.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009905.mm.1 |
NR_040314.1 |
Refseq |