Assay Details
Gene Symbol: A730056A06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040324.1 | 2 - 3 | 334 | 104 | ||
GenBank mRNA | BF470505.1 | - | 2 - 3 | 334 | 104 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 30075 |
Gm30075 | -75.1kb | Sense | Chr. 7 - 73201863 - 73238687 on NCBI Build GRCm38 | |
Downstream | repulsive guidance molecule family member A | Rgma |
BC059072, C230063O06 |
43.0kb | Anti-sense | Chr. 7 - 73375520 - 73419899 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2297300 | AK053954.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2297300 | BG065661.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2297300 | C76391.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2297300 | AI661470.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 457981 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 758570 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 842900 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 422620 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 443740 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 592668 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539172 | BC059104.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 145310 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 68394 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 462533 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 889612 | AK076537.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 624160 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 816110 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 286496 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 200876 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355748 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 120805 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 683062 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 173787 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 644866 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 203927 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 674481 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 868960 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 839780 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 962516 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 839778 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 869909 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 214448 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 125757 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559251 | AK010964.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559251 | AK168448.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559251 | AK216703.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559251 | AK218501.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559251 | BC006680.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559251 | BC008661.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559251 | BC021837.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559251 | BC025894.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559251 | BC036303.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559251 | BC094012.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559251 | D50527.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559251 | M81744.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559251 | NM_019639.4 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559251 | XM_001471699.1 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559251 | XM_001479832.1 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559251 | XM_001479835.1 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 86329 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 88737 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 109877 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 79739 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 22275 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145799 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2297300 | AK053954.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2297300 | BG065661.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2297300 | C76391.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2297300 | AI661470.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06240077 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01547428 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06240114 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06240095 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06240138 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06240003 | - | - |
Whole Mouse Genome G4122A | A_51_P380778 | AK053954.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P380778 | AK043101.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114083 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06240003 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06240077 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06240114 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228430 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228479 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05931549 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228329 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228384 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06536384 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228306 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228523 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228375 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228372 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228326 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228519 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228544 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228557 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228426 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228503 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228439 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228549 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06180982 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228550 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228521 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228371 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05949803 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228332 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228438 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06271342 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06180984 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228370 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05953782 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228515 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228367 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06378545 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06536382 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228307 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05949810 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06315414 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228484 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06180976 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228369 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228485 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06271918 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228558 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228376 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06368390 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228553 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228387 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06536379 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05931553 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06244428 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06126643 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228302 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228482 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228322 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06536385 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06599243 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228437 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228443 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228541 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06271923 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228334 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228304 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228510 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06315411 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228327 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228489 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228556 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228311 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228301 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06214349 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228517 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05929171 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228319 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228423 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228385 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228365 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228508 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228422 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228363 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228435 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228548 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228335 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228478 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228539 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05949808 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228441 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228530 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228374 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228309 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228513 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228546 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228543 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228554 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228551 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228320 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228542 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06235384 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06126644 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228533 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228324 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06180980 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228505 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06545473 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25228520 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1455415_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1455415_at | - | - |
Mouse Genome 430 2.0 Array | 1455415_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700029622.mm.1 | NR_040324.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700029622.mm.1 | NR_040324.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700029622.mm.1 | TC0700003492.mm.1 | MTA TC |
Mouse Expression Set 430 | 1455415_PM_at | - | - |
Mouse Expression Set 430 | 1455415_at | - | - |
Murine Genome U74v2 Set | 137652_at | - | - |
Murine Genome U74v2 Set | 137652_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455415_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455415_at | - | - |
