Assay Details
| Gene Symbol: AI646519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040330.1 | 1 - 2 | 440 | 73 | ||
| GenBank mRNA | AB080658.1 | - | 1 - 2 | 439 | 73 | |
| AI430246.1 | - | 1 - 2 | 56 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
NK2 homeobox 2, opposite strand |
Nkx2-2os |
Gm14112, Nkx2-2as, Nkx2.2AS, OTTMUSG00000015691 |
-29.9kb | Anti-sense | Chr. 2 - 147184083 - 147331681 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA A530006G24 gene | A530006G24Rik | Gm1003 | 347.9kb | Anti-sense | Chr. 2 - 147710693 - 147717632 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

AI646519
Genomic Map