Assay Details
Gene Symbol: A330093E20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040342.1 | 1 - 2 | 123 | 90 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
potassium intermediate/small conductance calcium-activated channel, subfamily N, member 2 |
Kcnn2 |
KCa2.2, SK2, SKCA2, fri |
-358.1kb | Anti-sense | Chr. 18 - 45268764 - 45685887 on NCBI Build GRCm38 |
Downstream | tripartite motif-containing 36 | Trim36 |
D18Wsu100e, haprin |
120.0kb | Sense | Chr. 18 - 46165300 - 46212607 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1800001374.rn
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
