Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1458264_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1458264_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1458264_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0800004253.mm.1 |
NR_040698.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004253.mm.1 |
NR_040698.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004253.mm.1 |
TC0800000632.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800004252.mm.1 |
NR_040698.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004252.mm.1 |
NR_040698.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004252.mm.1 |
TC0800000632.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800004254.mm.1 |
NR_040698.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004254.mm.1 |
NR_040698.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004254.mm.1 |
TC0800000632.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1458264_at |
- |
- |
Mouse Expression Set 430 |
1458264_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1458264_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1458264_at |
- |
- |