Assay Details
Gene Symbol: 4930546K05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040754.1 | 6 - 7 | 1844 | 108 | ||
GenBank mRNA | AK019746.1 | - | 3 - 4 | 395 | 108 | |
AK133167.1 | - | 6 - 7 | 1845 | 108 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 39321 |
Gm39321 | -36.8kb | Anti-sense | Chr. 9 - 42142092 - 42145394 on NCBI Build GRCm38 | |
Downstream |
sterol-C5-desaturase (fungal ERG3, delta-5-desaturase) homolog (S. cerevisae) |
Sc5d |
A830037K02, A830073K23Rik, Sc5dl |
65.3kb | Sense | Chr. 9 - 42251592 - 42264300 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 585820 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 858012 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 507000 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 476420 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 959988 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593223 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 43856 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170657 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01914315 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170657 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06763513 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110468 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170657 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07775407 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453938 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453841 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26454022 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453851 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453953 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453988 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453854 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453979 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07500210 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453956 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453944 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453962 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453984 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453999 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453941 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08121845 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07820074 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26454012 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453955 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453945 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26454007 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453957 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07558874 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453998 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453769 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453771 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453997 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07558869 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453777 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453839 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453783 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453781 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453960 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26454015 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07558868 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453986 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453958 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453843 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26454008 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07542520 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453971 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453949 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07470721 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26454001 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26454021 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453940 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07488359 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26454020 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07470719 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453933 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453936 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26454011 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453977 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453838 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26454027 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453995 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453947 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453935 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07535568 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26454030 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08017811 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453853 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453966 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453987 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07500219 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453973 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26454000 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453991 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453845 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26454023 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07488352 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453969 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07792805 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26454034 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07801662 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453779 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453776 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07479723 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453982 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453994 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453964 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26454024 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26453774 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26454004 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07792813 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07908863 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR0900017498.mm.1 | NR_040754.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900017498.mm.1 | NR_040754.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900017498.mm.1 | TC0900002114.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900017500.mm.1 | NR_040754.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900017500.mm.1 | NR_040754.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900017500.mm.1 | TC0900002114.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900017499.mm.1 | NR_040754.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900017499.mm.1 | NR_040754.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900017499.mm.1 | TC0900002114.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 103830452 | - | - |
