Assay Details
Gene Symbol: 4930519F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040763.1 | 2 - 3 | 458 | 107 | ||
GenBank mRNA | AK076865.1 | - | 2 - 3 | 461 | 107 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | SRY (sex determining region Y)-box 14 | Sox14 | -147.5kb | Anti-sense | Chr. 9 - 99874106 - 99876170 on NCBI Build GRCm38 | |
Downstream | predicted gene 28167 | Gm28167 | 236.5kb | Anti-sense | Chr. 9 - 100271212 - 100323517 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047763 | AK076865.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047763 | CF105892.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 383200 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 187890 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 156743 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 178006 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047763 | AK076865.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047763 | CF105892.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06899324 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06899324 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767309 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767420 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767304 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767388 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767337 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767397 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767345 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767424 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767266 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767274 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08044512 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767396 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767414 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767349 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767361 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767283 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767253 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767330 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767305 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767390 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767320 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07831348 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767376 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767307 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08069840 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767327 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767265 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767300 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767294 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767343 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767311 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767268 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767370 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767281 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767393 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767332 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767254 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767377 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767380 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767413 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767358 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767341 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767280 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767398 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767273 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767333 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767335 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08035785 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767286 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07798436 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767323 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767418 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767292 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08035777 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767322 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767423 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767276 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767411 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767400 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767288 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767298 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767303 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767347 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767328 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767392 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767391 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767363 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767317 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767384 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767385 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767372 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767336 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767263 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767279 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767342 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767425 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08069769 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767374 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767421 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767326 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767290 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767302 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767299 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767334 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767339 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767416 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767287 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767395 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26767314 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07503711 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR0900009840.mm.1 | NR_040763.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900009840.mm.1 | NR_040763.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900009840.mm.1 | TC0900001257.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900009841.mm.1 | NR_040763.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900009841.mm.1 | NR_040763.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900009841.mm.1 | TC0900001257.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 700486 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_2479556 | - | - |
