Assay Details
Gene Symbol: Gm5441 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_044987.1 | 2 - 3 | 304 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 10421 | Gm10421 | ENSMUSG00000072792 | -83.3kb | Sense | Chr. 12 - 117144523 - 117151269 on NCBI Build GRCm38 |
Downstream |
predicted gene, 39559 |
Gm39559 | 85.4kb | Anti-sense | Chr. 12 - 117345562 - 117347995 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 417966 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 612801 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 670369 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441274 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 533000 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 48050 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 53258 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 383770 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610823 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 252685 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 476093 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 315154 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 213095 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 34078 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 51003 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 842 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 547425 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 457572 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 567511 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546322 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 380310 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 691373 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 425304 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 50693 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 97228 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 60900 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009196 | AK039060.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07764074 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07764011 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07764041 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07764011 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772991 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773132 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773162 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09995203 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773066 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772964 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10067548 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772907 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772970 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10177287 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772905 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773151 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773144 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772913 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773169 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773117 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773137 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772911 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773126 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772981 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10368447 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773128 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773171 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10167367 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10332045 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10148715 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772983 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09786328 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772977 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09928524 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10334280 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09881781 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10334277 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773054 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10148716 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773064 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09881786 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772954 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773156 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772917 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09876603 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773121 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10390367 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773035 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773148 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773166 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773123 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773153 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772961 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773120 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773067 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10334274 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773045 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772952 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772988 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773049 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773043 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772974 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773164 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773046 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773038 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773129 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773044 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10381280 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772972 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773138 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773052 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773158 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772982 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773032 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773062 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773143 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772967 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773135 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10409228 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773033 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773159 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773041 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10187496 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772959 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09789424 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10170589 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773051 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773119 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773056 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09907521 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772985 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09907530 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773127 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772969 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772975 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773160 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28772910 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09789435 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28773145 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR1200018626.mm.1 | NR_044987.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200018626.mm.1 | NR_044987.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200018626.mm.1 | TC1200002504.mm.1 | MTA TC |
