Assay Details
| Gene Symbol: 4933411G06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045158.1 | 1 | 624 | 81 | ||
| GenBank mRNA | AK016770.1 | - | 1 | 625 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
regulatory factor X, 6 |
Rfx6 |
4930572O07Rik, Rfxdc1 |
-25.7kb | Anti-sense | Chr. 10 - 51677752 - 51730432 on NCBI Build GRCm38 |
| Downstream | vestigial like 2 homolog (Drosophila) | Vgll2 |
C130057C21Rik, VITO-1, Vito1, vgl-2 |
265.3kb | Anti-sense | Chr. 10 - 52022502 - 52028471 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4933411G06Rik
Genomic Map