Assay Details
Gene Symbol: 4933427I22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045160.1 | 1 - 2 | 81 | 95 | ||
GenBank mRNA | BE687684.1 | - | 1 - 2 | 81 | 95 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | paired box 7 | Pax7 | Pax-7 | -99.2kb | Sense | Chr. 4 - 139737059 - 139833026 on NCBI Build GRCm38 |
Downstream | kelch domain containing 7A | Klhdc7a | B230308G19Rik | 29.1kb | Sense | Chr. 4 - 139962173 - 139968026 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
