Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100026003.mm.1 |
NR_045273.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026003.mm.1 |
NR_045273.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026003.mm.1 |
TC1100002864.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100026004.mm.1 |
NR_045273.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026004.mm.1 |
NR_045273.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026004.mm.1 |
TC1100002864.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100026005.mm.1 |
NR_045273.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026005.mm.1 |
NR_045273.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026005.mm.1 |
TC1100002864.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100026001.mm.1 |
NR_045273.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026001.mm.1 |
NR_045273.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026001.mm.1 |
TC1100002864.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100026002.mm.1 |
NR_045273.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026002.mm.1 |
NR_045273.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026002.mm.1 |
TC1100002864.mm.1 |
MTA TC |