Assay Details
Gene Symbol: B230214G05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045281.1 | 1 - 2 | 167 | 80 | ||
GenBank mRNA | AK045598.1 | - | 1 - 2 | 168 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 6393 | Mir6393 |
Gm27424, mmu-mir-6393 |
-518.9kb | Anti-sense | Chr. 15 - 87795850 - 87795943 on NCBI Build GRCm38 |
Downstream |
zinc finger, DHHC domain containing 25 |
Zdhhc25 | 1700030J15Rik | 235.7kb | Sense | Chr. 15 - 88600302 - 88601669 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 56048 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 213838 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 106955 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 433840 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 215481 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478276 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 709049 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 795957 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 665890 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 454653 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 893748 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 279353 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 846179 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 943792 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 604055 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 750167 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 688823 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 379479 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 72380 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 16090 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08452585 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08452659 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08452628 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08452604 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103041 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08452628 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12396546 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418700 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418710 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418822 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418899 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418890 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418904 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12051770 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418681 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418887 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418943 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12055743 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418932 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418959 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418720 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12288942 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418871 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418973 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418946 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12181172 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418817 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418713 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418965 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418939 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418723 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12396095 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418692 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418697 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418722 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418699 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418955 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418714 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418938 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418702 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418875 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418684 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12232058 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12363278 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12055726 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418878 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418690 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418696 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11979233 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12055749 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418884 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12181163 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12181155 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418879 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418694 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418823 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12067850 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418941 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418704 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12149414 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418707 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418873 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418934 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418963 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418814 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418962 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418935 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418954 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418820 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418683 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12232051 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418712 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418810 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418937 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418688 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418882 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418685 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418825 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30418894 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR1500006640.mm.1 | NR_045281.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500006640.mm.1 | NR_045281.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500006640.mm.1 | TC1500000848.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 104060546 | - | - |
