Assay Details
Gene Symbol: Rptoros | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045313.1 | 1 - 2 | 81 | 103 | ||
GenBank mRNA | AK086908.1 | - | 1 - 2 | 82 | 103 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 11762 | Gm11762 | OTTMUSG00000004029 | -256.8kb | Anti-sense | Chr. 11 - 119548630 - 119569046 on NCBI Build GRCm38 |
Downstream | charged multivesicular body protein 6 | Chmp6 | 2400004G01Rik | 79.1kb | Anti-sense | Chr. 11 - 119913446 - 119919552 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
