Assay Details
| Gene Symbol: Gm19705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045324.1 | 1 - 2 | 80 | 99 | ||
| GenBank mRNA | BE447944.1 | - | 1 - 2 | 80 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 281 | Zfp281 | Znf281 | -53.0kb | Sense | Chr. 1 - 136624901 - 136630391 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 16880 |
Gm16880 | 7.4kb | Sense | Chr. 1 - 136696211 - 136725728 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm19705
Genomic Map