Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500005273.mm.1 |
TC0500000597.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500005273.mm.1 |
NR_045411.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005273.mm.1 |
NR_045411.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005273.mm.1 |
NR_045412.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005273.mm.1 |
NR_045412.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005273.mm.1 |
NR_045413.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005273.mm.1 |
NR_045413.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005282.mm.1 |
TC0500000597.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500005282.mm.1 |
NR_045413.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005282.mm.1 |
NR_045413.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005283.mm.1 |
TC0500000597.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500005283.mm.1 |
NR_045413.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005283.mm.1 |
NR_045413.1 |
Refseq |