Assay Details
Gene Symbol: 4930558G05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045441.1 | 3 - 4 | 321 | 88 | ||
GenBank mRNA | AK016174.1 | - | 3 - 4 | 324 | 88 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 4916 | Gm4916 |
EG237030, Rbm31x |
-689.1kb | Sense | Chr. X - 128375395 - 128377169 on NCBI Build GRCm38 |
Downstream | diaphanous homolog 2 (Drosophila) | Diap2 |
Dia3, Diaph2, Drf2, E430022I22Rik |
682.5kb | Sense | Chr. X - 129749668 - 130465834 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
