Assay Details
| Gene Symbol: AI507597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033566.1 | 2 - 3 | 213 | 93 | ||
| GenBank mRNA | AK131719.1 | - | 2 - 3 | 214 | 93 | |
| AK131785.1 | - | 2 - 3 | 214 | 93 | ||
| Gene Symbol: Tmem82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BU961312.1 | - | 2 - 3 | 232 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | filamin binding LIM protein 1 | Fblim1 |
2410043F08Rik, Cal, Fblp1, Gt10, migfilin, migfilin(s) |
-8.5kb | Anti-sense | Chr. 4 - 141576062 - 141606052 on NCBI Build GRCm38 |
| Downstream |
solute carrier family 25, member 34 |
Slc25a34 | Gm1369 | 3.2kb | Anti-sense | Chr. 4 - 141618824 - 141623993 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

AI507597
Genomic Map