Array Name |
Probe IDs |
Targets |
Target Type |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
58839 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
576685 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
301589 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
457712 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
630179 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
840904 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
507295 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
747621 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
100342 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
396728 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
865578 |
AK039944.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
865578 |
AK050306.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
865578 |
AK053715.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
865578 |
AK084697.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
865578 |
AK086395.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
865578 |
AK173211.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
865578 |
NM_178912.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
105185 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
89451 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
20581 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P177121 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07158187 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02193727 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P177121 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P177121 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364575 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364719 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364729 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364682 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364690 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08308325 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364713 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364694 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08814079 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364621 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364722 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364570 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364715 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364728 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364727 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364622 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08309361 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364717 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364687 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364742 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364743 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08934221 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364580 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364586 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08955534 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08671727 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364568 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364567 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08309366 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364576 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364723 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364680 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364749 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364625 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364695 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364739 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364684 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364744 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364747 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08930611 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364572 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364623 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364619 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08228810 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364746 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364737 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08659186 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364725 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08671726 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364712 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27364716 |
- |
- |