Assay Details
| Gene Symbol: 5430428K19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045426.1 | 3 | 817 | 77 | ||
| GenBank mRNA | AK017356.1 | - | 3 | 818 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | shroom family member 4 | Shroom4 |
D430043L16Rik, Gm724, Shrm4 |
Anti-sense | Chr. X - 6399938 - 6637452 on NCBI Build GRCm38 | |
| Downstream | diacylglycerol kinase kappa | Dgkk | Gm360 | 326.2kb | Anti-sense | Chr. X - 6873484 - 6948363 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5430428K19Rik
Genomic Map