Assay Details
| Gene Symbol: Crxos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205274.1 | NP_001192203.1 | 3 | 1512 | 100 | |
| NR_038092.1 | 5 | 1626 | 100 | |||
| GenBank mRNA | AK149251.1 | - | 5 | 1626 | 100 | |
| AK149256.1 | - | 5 | 1627 | 100 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Crxos
Genomic Map