Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1420181_PM_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1456942_PM_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1456942_PM_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1456942_x_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
668339_PM_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
668339_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0X00017377.mm.1 |
NM_001177365.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017377.mm.1 |
NM_001177365.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017377.mm.1 |
TC0X00002756.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00017376.mm.1 |
NM_001177365.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017376.mm.1 |
NM_001177365.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017376.mm.1 |
TC0X00002756.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00017374.mm.1 |
NM_001177365.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017374.mm.1 |
NM_001177365.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017374.mm.1 |
TC0X00002756.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00017375.mm.1 |
NM_001177365.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017375.mm.1 |
NM_001177365.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017375.mm.1 |
TC0X00002756.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1420181_PM_at |
- |
- |
Mouse Expression Set 430 |
1456942_x_at |
- |
- |
Mouse Expression Set 430 |
1420181_at |
- |
- |
Mouse Expression Set 430 |
1456942_PM_x_at |
- |
- |
Murine Genome U74v2 Set |
141044_PM_f_at |
- |
- |
Murine Genome U74v2 Set |
141044_f_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1449762_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1456942_x_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1449762_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1456942_PM_x_at |
- |
- |