Assay Details
| Gene Symbol: 4933439C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015585.2 | 3 - 4 | 1430 | 55 | ||
| GenBank mRNA | AK011602.1 | - | 3 - 4 | 963 | 55 | |
| AK017115.1 | - | 3 - 4 | 1431 | 55 | ||
| AK030920.1 | - | 1458 | 55 | |||
| AK163429.1 | - | 3 - 4 | 1426 | 55 | ||
| AK169756.1 | - | 3 - 4 | 1460 | 55 | ||
| BC065402.1 | - | 2 - 3 | 136 | 55 | ||
| BC099591.1 | - | 3 - 4 | 677 | 55 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger with KRAB and SCAN domains 17 | Zkscan17 |
Nizp1, Zfp496, Znf496 |
-1.2kb | Anti-sense | Chr. 11 - 59485520 - 59506640 on NCBI Build GRCm38 |
| Downstream |
NLR family, pyrin domain containing 3 |
Nlrp3 |
AGTAVPRL, AII/AVP, Cias1, FCAS, FCU, MWS, Mmig1, NALP3, Pypaf1 |
31.8kb | Sense | Chr. 11 - 59541569 - 59566956 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4933439C10Rik
Genomic Map