Assay Details
| Gene Symbol: Gm6607 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033622.2 | 1 | 116 | 120 | ||
| GenBank mRNA | BC011413.1 | - | 1 | 118 | 120 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 810 | Zfp810 | BC005471 | -22.5kb | Sense | Chr. 9 - 22276737 - 22307640 on NCBI Build GRCm38 |
| Downstream |
anillin, actin binding protein |
Anln |
1110037A17Rik, 2900037I21Rik, C78101, Scraps |
524.0 | Sense | Chr. 9 - 22331214 - 22389696 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm6607
Genomic Map