Assay Details
| Gene Symbol: Mir3971 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_039553.1 | 1 | 43 | 77 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 43, member 2 |
Slc43a2 |
7630402D21Rik, BC042513, Lat4 |
Sense | Chr. 11 - 75530282 - 75577575 on NCBI Build GRCm38 | |
| Downstream |
phosphatidylinositol transfer protein, alpha |
Pitpna |
Pitpn, vb, vibrator |
36.6kb | Sense | Chr. 11 - 75588108 - 75628778 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir3971
Genomic Map