Assay Details
| Gene Symbol: Mir5100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_039559.1 | 1 | 34 | 56 | ||
| GenBank mRNA | JF446399.1 | - | 1 | 34 | 56 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitochondrial elongation factor 2 | Mief2 |
AI482195, Gm11, Smcr7 |
Sense | Chr. 11 - 60728398 - 60732951 on NCBI Build GRCm38 | |
| Downstream | topoisomerase (DNA) III alpha | Top3a | 10.7kb | Anti-sense | Chr. 11 - 60739473 - 60777365 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir5100
Genomic Map